Skip to Content
Merck
All Photos(1)

Key Documents

Y0001137

Clobetasone butyrate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Clobetasone butyrate, 21-Chloro-9-fluoro-17-hydroxy-16-methylpregna-1,4-diene-3,11,20-trione 17-butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32ClFO5
CAS Number:
Molecular Weight:
478.98
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

clobetasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)C(=O)C[C@]12C)C(=O)CCl

InChI

1S/C26H32ClFO5/c1-5-6-22(32)33-26(21(31)14-27)15(2)11-19-18-8-7-16-12-17(29)9-10-23(16,3)25(18,28)20(30)13-24(19,26)4/h9-10,12,15,18-19H,5-8,11,13-14H2,1-4H3/t15-,18-,19-,23-,24-,25-,26-/m0/s1

InChI key

FBRAWBYQGRLCEK-AVVSTMBFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clobetasone butyrate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Equal efficacy of topical tacrolimus and clobetasone butyrate in pemphigus foliaceus.
Stuart N Cohen et al.
International journal of dermatology, 45(11), 1379-1379 (2006-11-02)
Salina Tsai et al.
International journal of dermatology, 44(5), 406-407 (2005-05-05)
A 45-year-old white woman presented with several years' history of firm, shiny papules on the lateral hands with slight extension to the dorsal fingers. The lesions first appeared between the index fingers and thumbs on both hands. They gradually increased
Divya Mathews et al.
Ophthalmic plastic and reconstructive surgery, 26(3), 206-207 (2010-05-22)
A 32-year-old woman presented with cystic eyelid lesions accompanied by foul-smelling discharge. Excision biopsy showed that she had Darier disease, and she was referred to dermatology for further management. Despite clinically indolent-looking lesions, biopsy and histology allowed the diagnosis of
Disseminated superficial actinic porokeratosis of the scalp.
K K Naineni et al.
Clinical and experimental dermatology, 32(2), 222-223 (2006-12-02)
Concomitant sensitivity to sodium metabisulfite and clobetasone butyrate in Trimovate cream.
D A Harrison et al.
Contact dermatitis, 46(5), 310-310 (2002-06-27)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service