Skip to Content
Merck
All Photos(1)

Key Documents

Y0000667

Human coagulation factor VII concentrate BRP

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Factor VII from human plasma, Proconvertin

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
€133.00

€133.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 EA
€133.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€133.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Human coagulation factor VII concentrate BRP EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chandrakumar Balaratnasingam et al.
Experimental eye research, 121, 11-22 (2014-02-25)
Retinal ganglion cell (RGC) axonal structure and function in the optic nerve head (ONH) is predominantly supported by astrocytes and capillaries. There is good experimental evidence to demonstrate that RGC axons are perturbed in a non-uniform manner following ONH injury
Hossein Ayatollahi et al.
Phytotherapy research : PTR, 28(4), 539-543 (2013-06-05)
Saffron showed some effects on blood coagulation and platelet aggregation in in vitro and in vivo studies. In a clinical trial with a limited number volunteers, saffron tablets influenced on bleeding time. In this study, the effect of saffron on
ACP Journal Club. 4F-PCC was noninferior to plasma in patients with acute major bleeding who needed urgent VKA reversal.
Eddy Lang
Annals of internal medicine, 160(8), JC6-JC6 (2014-04-16)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service