Skip to Content
Merck
All Photos(1)

Key Documents

W460822

Sigma-Aldrich

Guaiacol isobutyrate

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O3
CAS Number:
Molecular Weight:
194.23
FEMA Number:
4608
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.945
NACRES:
NA.21
grade:
FG
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

application(s)

flavors and fragrances

food allergen

no known allergens

Organoleptic

nutty; fruity

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

201.0 °F

Flash Point(C)

93.89 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service