Skip to Content
Merck
All Photos(3)

Key Documents

W455410

Sigma-Aldrich

cis-6-Nonenyl acetate

≥95%, stabilized, FG

Synonym(s):

(Z)-6-nonen-1-yl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

W455410-SAMPLE-K
€47.10
25 G
€93.80
100 G
€282.00

€47.10


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W455410-SAMPLE-K
€47.10
25 G
€93.80
100 G
€282.00

About This Item

Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
FEMA Number:
4554
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.673
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
food allergen:
no known allergens

€47.10


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

FG
Fragrance grade
Halal
Kosher

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

contains

synthetic α-tocopherol as stabilizer

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; melon; sweet

SMILES string

O(CCCCC\C=C/CC)C(=O)C

InChI

1S/C11H20O2/c1-3-4-5-6-7-8-9-10-13-11(2)12/h4-5H,3,6-10H2,1-2H3/b5-4-

InChI key

DFRXMRZQBMFKMI-PLNGDYQASA-N

Related Categories

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service