Skip to Content
Merck
All Photos(1)

Key Documents

W379312

Sigma-Aldrich

D-Ribose

natural sourced

Synonym(s):

D-Ribofuranose

Sign Into View Organizational & Contract Pricing

Select a Size

W379312-SAMPLE-K
€52.30
100 G
€122.00
5 KG
€594.00
1 KG
€717.00

€52.30


Estimated to ship on25 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W379312-SAMPLE-K
€52.30
100 G
€122.00
5 KG
€594.00
1 KG
€717.00

About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
FEMA Number:
3793
EC Number:
UNSPSC Code:
12352201
NACRES:
NA.21
Organoleptic:
odorless
grade:
Halal
Kosher
biological source:
corn
food allergen:
no known allergens

€52.30


Estimated to ship on25 March 2025


Request a Bulk OrderRequest more information

biological source

corn

Quality Level

grade

Halal
Kosher

application(s)

flavors and fragrances

food allergen

no known allergens

Organoleptic

odorless

SMILES string

O1C(C(C(C1CO)O)O)O

InChI

1S/C5H10O5/c6-1-2-3(7)4(8)5(9)10-2/h2-9H,1H2

InChI key

HMFHBZSHGGEWLO-UHFFFAOYSA-N

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service