Skip to Content
Merck
All Photos(1)

Key Documents

W355220

Sigma-Aldrich

2-hexyl-2-cyclopenten-1-one

sum of isomers, stabilized

Synonym(s):

Isojasmone

Sign Into View Organizational & Contract Pricing

Select a Size

W355220-SAMPLE-K
€47.10
100 G
€282.00
1 KG
€1,130.00

€47.10


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W355220-SAMPLE-K
€47.10
100 G
€282.00
1 KG
€1,130.00

About This Item

Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
FEMA Number:
3552
MDL number:
UNSPSC Code:
12164502
Flavis number:
07.033
NACRES:
NA.21
grade:
Fragrance grade
Halal
Kosher
food allergen:
no known allergens

€47.10


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

contains

synthetic α-tocopherol as stabilizer

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

jasmine; floral; waxy

SMILES string

O=C1CCC=C1CCCCCC

InChI

1S/C11H18O/c1-2-3-4-5-7-10-8-6-9-11(10)12/h8H,2-7,9H2,1H3

InChI key

VGECIEOJXLMWGO-UHFFFAOYSA-N

Related Categories

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

237.2 °F

Flash Point(C)

114 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service