Skip to Content
Merck
All Photos(1)

Key Documents

VP002

SAFC

VirusExpress® 293 AAV Production Cells

Pharma Manufacturing

Synonym(s):

HEK293 cells for AAV production

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

shipped in

liquid nitrogen

storage temp.

−196°C

Related Categories

General description

VirusExpress® 293 AAV Production Cells are a clonal, suspension-adapted HEK293 derivative selected for fast growth, high transfection efficiency, and high-titer AAV production. VirusExpress 293 AAV Production Cells grow to high cell densities and are amenable to chemical-based transfection reagents commonly used in manufacturing (i.e., PEI, lipid-nanoparticle). A medium exchange step is not needed prior to transfection, and the cells can be transfected in larger volumes, enabling scale-up of AAV production. The cells are supplied in a chemically defined media formulation and can be thawed directly into EX-CELL® CD HEK293 Viral Vector Medium.

VirusExpress 293 AAV Production Cells are derived from the HEK293 cell line (ATCC CRL-1573) through single cell cloning. The parental HEK293 cell line was established by transformation of human embryonic kidney cells with sheared human adenovirus 5 DNA (Graham F.L. et al. 1977). A 4-kb adenoviral genome fragment is known to have integrated into chromosome 19 and encodes for E1A/E1B proteins. Our clonal derivative eliminates variability from the population while also being adapted to suspension, serum-free conditions. Suspension culture and chemically defined medium allow scalability into stirred tank bioreactors for yields that can meet commercial needs.

Packaging

This product is shipped in a liquid nitrogen dry vapor shipper. The liquid nitrogen dry vapor shipper is not disposable and should be returned after receipt.

Other Notes

The cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL. The cells were banked under cGMP conditions in chemically defined medium and 8% DMSO.

Legal Information

A commercial license must be taken prior to use of the cells, or any products or materials derived from the cells, in a manufacturing process. Please contact your local sales representative for more details.
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
VirusExpress is a registered trademark of Merck KGaA, Darmstadt, Germany

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Scaling up viral vector production using adherent cell culture systems is challenging. Learn how suspension cell culture systems benefit large-scale bioprocessing.

A step-by-step overview of suspension-based, transient transfection bioreactor process development and scaleup of lentivirus production.

To address scalability challenges of AAV manufacturing, we developed an HEK293 suspension cell line that can be used across many serotypes. Get the data in this article.

The current status and future manufacturing of AAV-based gene therapies is discussed in this podcast transcript, including how to streamline large-scale manufacturing.

See All

Related Content

A transfection-based solution to viral vector production using a suspension cell line, chemically defined medium, and a process with proven performance at scale.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service