Select a Size
€ 29,60
€ 233,00
€ 29,60
About This Item
Skip To
1 of 4
This Item | |||
|---|---|---|---|
| form liquid | form solution | form solution | form - |
| shipped in dry ice | shipped in - | shipped in - | shipped in dry ice |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| Quality Level 300 | Quality Level 100 | Quality Level 100 | Quality Level 200 |
General description
Features and Benefits
- 10 min rapid inactivation at 55°C.
- Compatible with common PCR reaction systems.
Storage Class
10 - Combustible liquids
wgk
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Erfahren Sie mehr über hochwertige Rohstoffe für die Entwicklung diagnostischer Assays und die IVD-Herstellung. Profitieren Sie von unseren Fachkenntnissen in Bezug auf Tests für die molekulare Diagnostik und Enzyme für molekulare Assays.
Explore high-quality raw materials for diagnostic assay development and IVD manufacturing. Leverage our expertise in molecular diagnostic testing and enzymes for molecular assays.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| UDGEC-500U | 04065267928135 |
| UDGEC-5000U | 04065267928128 |
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


