Skip to Content
Merck
All Photos(1)

Key Documents

TSS10

Supelco

Total Suspended Solids 10 mg/L Calibration Standard

certified reference material

Synonym(s):

TSS Reference Solution (10 mg/L)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€97.10
500 ML
€136.00

€97.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€97.10
500 ML
€136.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€97.10


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

CofA

certificate is enclosed in each package

manufacturer/tradename

RTC TSS10-100

concentration

in H2O

application(s)

environmental

format

single component solution

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Total Suspended Solids 10 mg/L Calibration Standard for R&D and Analytical use only.

Features and Benefits

  • Packed in High Density Polyethylene (HDPE) bottle
  • A certificate of analysis (CoA) is included with the product
  • Traceable to recognized national standards, and units of measurement recognized at the National Institute of Standards and Technology (NIST) and International System of units (SI)
  • Gravimetrically performed dilutions
  • Balance used to prepare the standard is calibrated to ISO 17025 requirements
  • Stable solution with long minimum shelf life

Other Notes

Values of analytes vary lot to lot.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service