Skip to Content
Merck
All Photos(1)

Documents

T3006

Supelco

Testosterone enanthate

analytical standard

Synonym(s):

(17β)-17-[(1-Oxoheptyl)oxy]androst-4-en-3-one, 17β-Hydroxy-4-androsten-3-one 17-enanthate, 4-Androsten-17β-ol-3-one 17-enanthate, NSC 17591, Testosterone 17β-heptanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H40O3
CAS Number:
Molecular Weight:
400.59
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C26H40O3/c1-4-5-6-7-8-24(28)29-23-12-11-21-20-10-9-18-17-19(27)13-15-25(18,2)22(20)14-16-26(21,23)3/h17,20-23H,4-16H2,1-3H3/t20-,21-,22-,23-,25-,26-/m0/s1

InChI key

VOCBWIIFXDYGNZ-IXKNJLPQSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

Testosterone enanthate is a testosterone ester, which is widely used in hormone replacement therapy.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative pharmacokinetics of testosterone enanthate and testosterone cyclohexanecarboxylate as assessed by serum and salivary testosterone levels in normal men.
Schurmeryer T and Nieschlag E.
International Journal of Andrology, 7(3), 181-187 (1984)
Andrology: Male Reproductive Health and Dysfunction (2010)
Hanna M King et al.
Psychoneuroendocrinology, 37(3), 396-409 (2011-08-09)
Increasing evidence in humans and other animals suggests that testosterone (T) plays an important role in modulating emotion. We previously reported that T treatment in rhesus monkeys undergoing chemically induced hypogonadism results in increased watching time of videos depicting fights
Shane Rogerson et al.
Journal of strength and conditioning research, 21(2), 354-361 (2007-05-29)
Use of testosterone enanthate has been shown to significantly increase strength within 6-12 weeks of administration (2, 9), however, it is unclear if the ergogenic benefits are evident in less than 6 weeks. Testosterone enanthate is classified as a prohibited
Séverine Trabado et al.
Fertility and sterility, 95(7), 2324-2329 (2011-05-04)
To evaluate the degree of E2 deficiency in male congenital hypogonadotropic hypogonadism (CHH), and its response to different hormonal treatments. Retrospective and prospective studies. Academic institution. Untreated or treated CHH, healthy men, untreated men with Klinefelter syndrome (KS). Serum sex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service