Skip to Content
Merck
All Photos(4)

Key Documents

T0200

Sigma-Aldrich

Tannic acid

Source: Chinese natural gall nuts

Synonym(s):

Gallotannin, Tannin

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€69.70
500 G
€133.00
1 KG
€238.00

€69.70


Estimated to ship on22 March 2025


Request a Bulk Order

Select a Size

Change View
250 G
€69.70
500 G
€133.00
1 KG
€238.00

About This Item

Empirical Formula (Hill Notation):
C76H52O46
CAS Number:
Molecular Weight:
1701.20
Beilstein:
8186396
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

€69.70


Estimated to ship on22 March 2025


Request a Bulk Order

Quality Level

autoignition temp.

980 °F

mp

218 °C (lit.)

functional group

ester
ether

SMILES string

Oc1cc(cc(O)c1O)C(=O)Oc2cc(cc(O)c2O)C(=O)OC[C@H]3O[C@@H](OC(=O)c4cc(O)c(O)c(OC(=O)c5cc(O)c(O)c(O)c5)c4)[C@H](OC(=O)c6cc(O)c(O)c(OC(=O)c7cc(O)c(O)c(O)c7)c6)[C@@H](OC(=O)c8cc(O)c(O)c(OC(=O)c9cc(O)c(O)c(O)c9)c8)[C@@H]3OC(=O)c%10cc(O)c(O)c(OC(=O)c%11cc(O)c(O)c(O)c%11)c%10

InChI

1S/C76H52O46/c77-32-1-22(2-33(78)53(32)92)67(103)113-47-16-27(11-42(87)58(47)97)66(102)112-21-52-63(119-72(108)28-12-43(88)59(98)48(17-28)114-68(104)23-3-34(79)54(93)35(80)4-23)64(120-73(109)29-13-44(89)60(99)49(18-29)115-69(105)24-5-36(81)55(94)37(82)6-24)65(121-74(110)30-14-45(90)61(100)50(19-30)116-70(106)25-7-38(83)56(95)39(84)8-25)76(118-52)122-75(111)31-15-46(91)62(101)51(20-31)117-71(107)26-9-40(85)57(96)41(86)10-26/h1-20,52,63-65,76-101H,21H2/t52-,63-,64+,65-,76+/m1/s1

InChI key

LRBQNJMCXXYXIU-PPKXGCFTSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

390.2 °F

Flash Point(C)

199 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yufei Yuan et al.
Journal of insect science (Online), 20(1) (2020-02-16)
Plant tannins, polyphenolic plant secondary metabolites are involved in important chemical defense processes in plants. In this study, tannic acid was used as the standard of plant tannins to determine the effects on nutritional indices and activities of glutathione S-transferases
Claudia Scattino et al.
Food chemistry, 163, 51-60 (2014-06-11)
In the present study the possibility of enhancing phenolic compound contents in peaches and nectarines by post-harvest irradiation with UV-B was assessed. Fruits of 'Suncrest' and 'Babygold 7' peach and 'Big Top' nectarine cultivars were irradiated with UV-B for 12
Anna Müller et al.
BMC plant biology, 15, 165-165 (2015-07-01)
Chrysomela populi (poplar leaf beetle) is a common herbivore in poplar plantations whose infestation causes major economic losses. Because plant volatiles act as infochemicals, we tested whether isoprene, the main volatile organic compound (VOC) produced by poplars (Populus x canescens)
Sylvain Godreuil et al.
PloS one, 9(8), e105441-e105441 (2014-08-28)
Multidrug resistance, which is acquired by both Gram-positive and Gram-negative bacteria, causes infections that are associated with significant morbidity and mortality in many clinical settings around the world. Because of the rapidly increasing incidence of pathogens that have become resistant
Dmitri Kravtsov et al.
American journal of physiology. Gastrointestinal and liver physiology, 307(10), G992-G1001 (2014-09-27)
Microvillus inclusion disease (MVID) is an autosomal recessive condition resulting in intractable secretory diarrhea in newborns due to loss-of-function mutations in myosin Vb (Myo5b). Previous work suggested that the apical recycling endosomal (ARE) compartment is the primary location for phosphoinositide-dependent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service