Skip to Content
Merck
All Photos(1)

Key Documents

SRA1

Sigma-Aldrich

Anisaldehyde solution

≥97.5% purity (GC), liquid

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€146.00

€146.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
€146.00

About This Item

MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€146.00


Estimated to ship on30 May 2025


Request a Bulk Order

Product Name

Anisaldehyde solution,

description

Spray reagent

Quality Level

Assay

≥97.5% (GC)

form

liquid

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[H]C(C1=CC=C(OC)C=C1)=O

InChI

1S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3

InChI key

ZRSNZINYAWTAHE-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Application

Excellent reagent for detecting a wide variety of natural products on TLC plates: steroids, prostaglandins, carbohydrates, phenols, glycosides, sapogenins, antibiotics, mycotoxins, antioxidants,[1] etc. A range of colors is produced on heating, adding a qualitative aspect to the detection process.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

52.0 °F - closed cup

Flash Point(C)

11.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a probe for monitoring HSV1-tk
reporter gene expression using chemical exchange
saturation transfer MRI
Annon Bar
Nature (2013)
Jork, H., et al.
Thin Layer Chromatography: Reagents and Detection Methods, 195-198 (1990)
Yun-Hai Tao et al.
Bioorganic & medicinal chemistry letters, 16(3), 592-595 (2005-11-18)
4-Hydroxybenzaldehyde (HBA) derivatives were examined as inhibitors for GABA transaminase (GABA-T) and succinic semialdehyde dehydrogenase (SSADH). Investigation of structure-activity relation revealed that a carbonyl group or an amino group as well as a hydroxy group at the para position of
Li Liu et al.
Bioorganic & medicinal chemistry letters, 22(1), 154-163 (2011-12-06)
The binding between the estrogen receptor α (ER-α) and a variety of compounds in traditional Chinese formulae, Si-Wu-Tang (SWT) series decoctions, was studied using a stably-transfected human breast cancer cell line (MVLN). In 38 compounds tested from SWT series decoctions
Chao-Bin Xue et al.
Bioorganic & medicinal chemistry, 15(5), 2006-2015 (2007-01-30)
Phenoloxidase (PO), also known as tyrosinase, is a key enzyme in insect development, responsible for catalyzing the hydroxylation of tyrosine into o-diphenols and the oxidation of o-diphenols into o-quinones. Inhibition of PO may provide a basis for novel environmentally friendly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service