Skip to Content
Merck
All Photos(1)

Documents

SMB01362

Sigma-Aldrich

Arginylphenylalanine

≥95% (HPLC)

Synonym(s):

Arg-phe, Arginyl-phenylalanine, RF, RF dipeptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N5O3
CAS Number:
Molecular Weight:
321.37
MDL number:
UNSPSC Code:
12352209
UNSPSC Code:
12352200
NACRES:
NA.26

Quality Level

Assay

≥95% (HPLC)

form

solid

color

white to beige

solubility

water: soluble

storage temp.

2-8°C

SMILES string

NC(NCCC[C@H](N)C(N[C@@H](CC1=CC=CC=C1)C(O)=O)=O)=N

InChI

1S/C15H23N5O3/c16-11(7-4-8-19-15(17)18)13(21)20-12(14(22)23)9-10-5-2-1-3-6-10/h1-3,5-6,11-12H,4,7-9,16H2,(H,20,21)(H,22,23)(H4,17,18,19)/t11-,12-/m0/s1

InChI key

PQBHGSGQZSOLIR-RYUDHWBXSA-N

General description

Arginylphenylalanine is a dipeptide of arginine and phenylalanine. Arg-phe is considered a secondary metabolite and potential biomarker of resulting from the consumption of certain foods, such as chicken or pigs.

Application

Arginylphenylalanine can be used in biochemical and metabolomics research applications

Features and Benefits

  • Suitable for Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service