Skip to Content
Merck
All Photos(1)

Key Documents

SMB00961

Sigma-Aldrich

Amitriptyline N-oxide

research grade

Synonym(s):

Amitriptylinoxide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€105.00

€105.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€105.00

About This Item

Empirical Formula (Hill Notation):
C20H23NO
CAS Number:
Molecular Weight:
293.40
UNSPSC Code:
12352119
NACRES:
NA.77

€105.00


Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Quality Level

Assay

≥95%

form

solid

storage condition

under inert gas

technique(s)

HPLC: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

[N+]([O-])(CCC=C1c2c(cccc2)CCc3c1cccc3)(C)C

InChI

1S/C20H23NO/c1-21(2,22)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20/h3-6,8-12H,7,13-15H2,1-2H3

InChI key

ZPMKQFOGINQDAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amitriptyline N-oxide is the N-oxidation metabolite of the antidepressant amitriptyline

Application

Metabolomics research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Altendorfer-Kroath et al.
Journal of neuroscience methods, 311, 394-401 (2018-09-30)
Assessment of drug concentration in the brain interstitial fluid (ISF) is crucial for development of brain active drugs, which are mainly small, lipophilic substances able to cross the blood-brain barrier (BBB). We aimed to compare the applicability of cerebral Open
Judith M Kampa et al.
Toxicology letters, 325, 43-50 (2020-02-25)
As a consequence of the detoxification process, drugs and drug related metabolites can accumulate in the liver, resulting in drug induced liver injury (DILI), which is the major cause for dose limitation. Amitriptyline, a commonly used tricyclic anti-depressant, is known
Ursula Breyer-Pfaff
Drug metabolism reviews, 36(3-4), 723-746 (2004-11-24)
Amitriptyline (AT), the most widely used tricyclic antidepressant, undergoes oxidative metabolism in the side chain with production of the secondary amine nortriptyline (NT), a primary amine, and the N-oxide amitriptylinoxide (AT-NO); in addition, direct conjugation leads to a quaternary ammonium-linked

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service