Skip to Content
Merck
All Photos(1)

Key Documents

SMB00939

Sigma-Aldrich

1-Methyladenosine

≥95% (HPLC)

Synonym(s):

1-Methyladenosine, N1-Methyladenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N5O4
CAS Number:
Molecular Weight:
281.27
MDL number:
UNSPSC Code:
12352123
NACRES:
NA.79
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

color

white

storage temp.

-10 to -25°C

SMILES string

CN1C=Nc2c(ncn2[C@@H]3O[C@@H](CO)[C@H](O)[C@@H]3O)C1=N

InChI

1S/C11H15N5O4/c1-15-3-14-10-6(9(15)12)13-4-16(10)11-8(19)7(18)5(2-17)20-11/h3-5,7-8,11-12,17-19H,2H2,1H3/t5-,7-,8-,11-/m1/s1

InChI key

GFYLSDSUCHVORB-IOSLPCCCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Methyladenosine is an RNA modification that arises through distinct processes, including enzymatic catalysis and the alkylation of RNA by specific agents.
1-Methyladenosine, also known as M1A, belongs to the class of organic compounds known as purine nucleosides. Purine nucleosides are compounds comprising a purine base attached to a ribosyl or deoxyribosyl moiety. 1-Methyladenosine is a strong basic compound (based on its pKa). Precise m6A mapping by m6A-CLIP/IP (briefly m6A-CLIP) revealed that a majority of m6A locates in the last exon of mRNAs in multiple tissues/cultured cells of mouse and human, and the m6A enrichment around stop codons is a coincidence that many stop codons locate round the start of last exons where m6A is truly enriched. The methylation of adenosine is directed by a large m6A methyltransferase complex containing METTL3 as the SAM-binding sub-unit. Insulin-like growth factor-2 mRNA-binding proteins 1, 2, and 3 (IGF2BP1–3) are reported as a novel class of m6A readers.

Application

1-Methyladenosine, a modified nucleoside produced during tRNA processing by methyltransferases, has garnered attention as a potential biomarker for early cancer detection due to its elevated urinary excretion in various cancer forms. Additionally, its presence can be monitored in serum. Moreover, 1-Methyladenosine levels rise during active rheumatoid arthritis, making it a promising indicator for this condition. It finds application in research pertaining to cancer, autoimmune diseases, metabolomics, biomarker discovery and drug discovery studies.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service