Skip to Content
Merck
All Photos(1)

Key Documents

S9144

Supelco

Starch assay reagent

for use with enzymatic assay kits SA20, STA20

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€176.00

€176.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
1 VIAL
€176.00

About This Item

UNSPSC Code:
12164500
NACRES:
NA.84

€176.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Quality Level

analyte chemical class(es)

sugars

technique(s)

photometry: suitable

application(s)

food and beverages
general analytical

compatibility

for use with Starch Assay Kits SA-20 and STA-20

storage temp.

2-8°C

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service