Skip to Content
Merck
All Photos(1)

Key Documents

S4797

Sigma-Aldrich

Sodium tartrate dibasic dihydrate

BioXtra, ≥99.0%

Synonym(s):

L-(+)-Tartaric acid disodium salt, Disodium tartrate dihydrate, Sodium tartrate dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€62.00
500 G
€218.00

€62.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
100 G
€62.00
500 G
€218.00

About This Item

Empirical Formula (Hill Notation):
C4H4Na2O6 · 2H2O
CAS Number:
Molecular Weight:
230.08
Beilstein:
6121732
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

€62.00


Estimated to ship on16 April 2025


Request a Bulk Order

product line

BioXtra

Quality Level

Assay

≥99.0%

form

powder or crystals

impurities

≤0.0005% Phosphorus (P)
≤0.005% Insoluble matter

pH

7.0-9 (20 °C, 50 g/L)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.0005%
sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.003%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.2Na.2H2O/c5-1(3(7)8)2(6)4(9)10;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;2*1H2/q;2*+1;;/p-2/t1-,2-;;;;/m1..../s1

InChI key

FGJLAJMGHXGFDE-DGFHWNFOSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Serena Duchi et al.
Materials (Basel, Switzerland), 12(4) (2019-02-23)
Cartilage defects and diseases remain major clinical issues in orthopaedics. Biomanufacturing is now a tangible option for the delivery of bioscaffolds capable of regenerating the deficient cartilage tissue. However, several limitations of in vitro and experimental animal models pose serious
Siavash Hassanpour et al.
PloS one, 9(9), e109078-e109078 (2014-10-03)
Adseverin (Ads), a member of the Gelsolin superfamily of actin binding proteins, regulates the actin cytoskeleton architecture by severing and capping existing filamentous actin (F-actin) strands and nucleating the assembly of new F-actin filaments. Ads has been implicated in cellular
Christian E Jacome-Galarza et al.
Nature, 568(7753), 541-545 (2019-04-12)
Osteoclasts are multinucleated giant cells that resorb bone, ensuring development and continuous remodelling of the skeleton and the bone marrow haematopoietic niche. Defective osteoclast activity leads to osteopetrosis and bone marrow failure1-9, whereas excess activity can contribute to bone loss

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service