Skip to Content
Merck
All Photos(3)

Documents

S4429

Sigma-Aldrich

Sulindac

meets USP testing specifications

Synonym(s):

(Z)-5-Fluoro-2-methyl-1-[p-(methylsulfinyl)benzylidene]indene-3-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17FO3S
CAS Number:
Molecular Weight:
356.41
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

application(s)

pharmaceutical (small molecule)

SMILES string

CC1=C(CC(O)=O)c2cc(F)ccc2\C1=C/c3ccc(cc3)S(C)=O

InChI

1S/C20H17FO3S/c1-12-17(9-13-3-6-15(7-4-13)25(2)24)16-8-5-14(21)10-19(16)18(12)11-20(22)23/h3-10H,11H2,1-2H3,(H,22,23)/b17-9-

InChI key

MLKXDPUZXIRXEP-MFOYZWKCSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nonsteroidal anti-inflammatory; preferential inhibitor of COX-1.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen X Skapek et al.
Pediatric blood & cancer, 60(7), 1108-1112 (2013-01-03)
Desmoid fibromatosis (desmoid tumor, DT) is a soft tissue neoplasm prone to recurrence despite complete surgical resection. Numerous small retrospective reports suggest that non-cytotoxic chemotherapy using tamoxifen and sulindac may be effective for DT. We evaluated the safety and efficacy
Caihua Zhu et al.
Stem cells (Dayton, Ohio), 30(10), 2065-2075 (2012-06-02)
Pharmacological targeting of breast cancer stem cells (CSCs) is highly promising for the treatment of breast cancer, as the small population of CSCs appears responsible for tumor initiation and progression and also for resistance to conventional treatment. Here we report
D E Duggan
Drug metabolism reviews, 12(2), 325-337 (1981-01-01)
Sulindac is a prodrug which, following absorption, rapidly attains a metabolic equilibrium with its active pharmacophore, the sulfide metabolite. At the level of the whole body, the reversible interconversion sulindac in equilibrium sulfide, and the differing distributional and excretory properties
N M Davies et al.
Clinical pharmacokinetics, 32(6), 437-459 (1997-06-01)
Sulindac is a nonsteroidal anti-inflammatory drug (NSAID) of the indene acetic acid class. The absorption of sulindac is rapid when given orally. Sulindac is reversibly metabolised to sulindac sulphide which has anti-inflammatory and analgesic properties and is irreversibly metabolised to
Johanna Wanngren et al.
The Journal of biological chemistry, 287(39), 32640-32650 (2012-08-02)
The γ-secretase complex is an appealing drug target when the therapeutic strategy is to alter amyloid-β peptide (Aβ) aggregation in Alzheimer disease. γ-Secretase is directly involved in Aβ formation and determines the pathogenic potential of Aβ by generating the aggregation-prone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service