Skip to Content
Merck

RTM19

Mineral Oil Rotational Viscosity Standard

viscosity 351.1 mPa.s (25.00 °C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

351.1 mPa.s(25.00 °C)

application(s)

cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care

format

mixture

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Mineral Oil Rotational Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
RTM19-500ML04061837681387