Skip to Content
Merck

Skip To

RTM12

Mineral Oil Rotational Viscosity Standard

viscosity 55.68 mPa.s (25.00 °C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
500 mL
Check Cart for Availability
€ 146,00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

€ 146,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

55.68 mPa.s(25.00 °C)

application(s)

cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care

format

mixture

General description

Mineral Oil Rotational Viscosity Standards (viscosity 55.68 mPa.s) are specifically formulated for the calibration and verification of rotational viscometers where operating temperature is controlled precisely.

Features and Benefits

  • Tested and certified at 25°C
  • Tamper-evident security packaging
  • 500mL quantity supplied in ready-to-use "test in pack" container with tamper evident cap provides absolute assurance of sample integrity
  • Full traceability to national standards
  • 1-year shelf life


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RTM4RTM8RTM9
format

mixture

format

mixture

format

mixture

format

mixture

application(s)

cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care

application(s)

cleaning products
cosmetics
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

viscosity

55.68 mPa.s(25.00 °C)

viscosity

2.144 mPa.s(25.00 °C)

viscosity

10.37 mPa.s(25.00 °C)

viscosity

14.50 mPa.s(25.00 °C)


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

158.0 °F

flash_point_c

70 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
RTM12-500ML04061837681318

Questions

Reviews

No rating value

Active Filters