Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
500 mL | Check Cart for Availability | € 524,00 |
About This Item
grade
certified reference material
Quality Segment
shelf life
limited shelf life, expiry date on the label
packaging
pkg of 500 mL
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
storage temp.
2-8°C
General description
Paragon ISO 17025 / ISO 17034 certified Relative Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C to 150°C.
1 of 1
This Item | |||
|---|---|---|---|
| description UKAS ISO/IEC17025 and ISO 17034 certified, Relative Density 20/4 0.7145 at 20 °C | description UKAS ISO/IEC17025 and ISO 17034 certified, Relative Density 20/4 0.6597 at 20 °C | description UKAS ISO/IEC17025 and ISO 17034 certified, Relative Density 20/4 0.7769 at 20 °C | description UKAS ISO/IEC17025 and ISO 17034 certified, Relative Density 20/4 0.8400 at 20 °C |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| application(s) cleaning products | application(s) cleaning products | application(s) cleaning products | application(s) cleaning products |
| manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| packaging pkg of 500 mL | packaging pkg of 500 mL | packaging pkg of 500 mL | packaging pkg of 500 mL |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
Still not finding the right product?
Explore all of our products under Relative Density Standard (20°C)
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
-4.0 °F
flash_point_c
-20 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




