Skip to Content
Merck

Skip To

RDD032

1-Octanesulfonic acid sodium salt

free-flowing, Redi-Dri, ≥98%

Synonym(s):

Octane-1-sulfonic acid sodium salt, Sodium 1-octanesulfonate, Sodium octane-1-sulfonate, Sodium octanesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Check Cart for Availability
€ 476,00
500 g
Check Cart for Availability
€ 1.860,00

About This Item

Linear Formula:
CH3(CH2)7SO3Na
CAS Number:
Molecular Weight:
216.27
UNSPSC Code:
12161902
NACRES:
NA.28
MDL number:

€ 476,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Redi-Dri

Quality Segment

assay

≥98%

quality

free-flowing

bp

>300 °C (lit.)

mp

>300 °C (lit.)

SMILES string

[Na+].CCCCCCCCS([O-])(=O)=O

InChI

1S/C8H18O3S.Na/c1-2-3-4-5-6-7-8-12(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1

InChI key

HRQDCDQDOPSGBR-UHFFFAOYSA-M

General description

Sodium octanesulfonate, or 1-Octanesulfonic acid sodium salt, is an anionic surfactant commonly used for HPLC analysis of peptides and proteins. It is hydrophilic in nature, with a hydrocarbon chain, sulphonic acid group, and the ability to allow ion exchange phenomenon due to the presence of sodium ions.

Application

1-Octanesulfonic acid sodium salt is for research use to
  • separate carboxylic acids using ion-exclusion chromatography (SEC) in the mobile phase for High-Performance Liquid Chromotography (HPLC) to separate organic molecules[1]
  • in preparation of water-soluble pillar[5]arenes[2]
  • develop graphene oxide-based composite (GOBC) for decontaminate of wastewater

Biochem/physiol Actions

1-Octanesulfonic acid sodium salt is hydrophilic in nature and has the ability to allow the ion exchange phenomenon due to the presence of sodium ions. For the formation of GOBC, it intercalates GO to form a nanoporous graphene that has a high adsorption capacity

Features and Benefits

  • Redi-Dri makes using hygroscopic chemicals safer and easier to use, dispense, and weigh out powders that commonly clump
  • Eliminates clumping by maintaining product moisture and quality attributes.
  • No caking makes it safer to use with no need to manually break up clumps with tools
  • Less waste - hydroscopic chemicals that normally clump and often are discarded

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC


Still not finding the right product?

Explore all of our products under 1-Octanesulfonic acid sodium salt


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters