Skip to Content
Merck
All Photos(1)

Documents

R8881

Sigma-Aldrich

Rhodamine B isothiocyanate–Dextran

Powder

Synonym(s):

RITC-Dextran

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

product name

Rhodamine B isothiocyanate–Dextran, average mol wt ~10,000

form

powder

Quality Level

mol wt

average mol wt ~10,000

extent of labeling

0.002-0.01 mol RITC per mol glucose

color

light pink to red
very dark purple

solubility

water: 10 mg/mL, clear, pink

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Rhodamine B is a thiol-reactive and photostable fluorescent dye. Rhodamine B isothiocyanate is an aminoxanthene that possesses carboxylic acid and isothiocyanate substituents on the pendant phenyl ring.

Application

Rhodamine B isothiocyanate (NE)-labeled dextran is often used as a fluorescent probe for studies of endocytotic activity and to detect the migrated of marked cells. It is also suitable for perfusion studies in animals. Rhodamine B isothiocyanate–Dextran has been used for in vivo zebrafish blood-brain barrier (BBB) dye diffusion assay.
Dextran labeled with rhodamine B isothiocyanate for possible use in perfusion studies in animals.

Biochem/physiol Actions

Rhodamine B stains the tissue in red-violet color and emits a bright red color when fluoresced. It is highly preferred for marking insects.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaotong Wu et al.
Journal of molecular cell biology, 10(6), 479-493 (2018-07-31)
Oocytes, the irreplaceable gametes for generating a new organism, are matured in the ovary of living female animals. It is unknown whether any genetic manipulations can be applied to immature oocytes inside the living ovaries. As a proof-of-concept, we here
Specific binding of a mutated fragment of Clostridium perfringens enterotoxin to endothelial claudin-5 and its modulation of cerebral vascular permeability
Zhuangbin Liao
Neuroscience (2016)
Jeremy Gingrich et al.
Chemosphere, 263, 128304-128304 (2020-11-07)
Gap junction intercellular communication (GJIC) is necessary for ovarian function, and it is temporospatially regulated during follicular development and ovulation. At outermost layer of the antral follicle, theca cells provide structural, steroidogenic, and vascular support. Inter- and extra-thecal GJIC is
Ruitao Su et al.
Science advances, 6(41) (2020-10-11)
Microfluidic devices fabricated via soft lithography have demonstrated compelling applications such as lab-on-a-chip diagnostics, DNA microarrays, and cell-based assays. These technologies could be further developed by directly integrating microfluidics with electronic sensors and curvilinear substrates as well as improved automation
Jiawei Sun et al.
Development (Cambridge, England), 145(19) (2018-08-24)
Maternal mRNAs and proteins dictate early embryonic development before zygotic genome activation. In the absence of transcription, elaborate control of maternal mRNA translation is of particular importance for oocyte maturation and early embryogenesis. By analyzing zebrafish ybx1 mutants with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service