Skip to Content
Merck
All Photos(3)

Key Documents

PPB023

Sigma-Aldrich

Tris buffer

Pre-set, pH 8.1, pHast Pack, powder

Synonym(s):

0.1M Tris

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
€94.90

€94.90


Estimated to ship on04 April 2025


Request a Bulk Order

Select a Size

Change View
20 PKG
€94.90

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

€94.90


Estimated to ship on04 April 2025


Request a Bulk Order

Quality Level

product line

pHast Pack

form

powder

storage condition

(Tightly closed. Dry)

impurities

≤5 ppm Heavy Metals (lead)

color

white

pH

8.1

cation traces

Fe: ≤10 ppm

foreign activity

DNase, none detected
Endonuclease, none detected
Exonuclease, none detected
NICKase, none detected
Protease, none detected
RNase, none detected

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Tris buffer, pH 8.1 is a general biological buffer commonly used in biochemistry and molecular biology. It is isotonic and non-toxic in nature due to the presence of tris base. Tris has a buffering capacity range of pH 7.0 - 9.0, with a pKa of 8.1 at 25°C.

Application

Intended for use in:
  • Cell culture (sterile filter recommended)
  • Western blot
  • Protein electrophoresis
  • DNA agarose electrophoresis as a component to make Tris acetate-EDTA (TAE) and Tris borate-EDTA (TBE) buffers

Features and Benefits

  • Ready-to-use for quick buffer prep
  • No measuring and adjusting pH
  • Saves time and minimizes effort in the lab
  • Biological tests: free of DNase, RNase, protease, and nickase
  • Chemical tests: Iron ≤10 ppm, lead ≤5 ppm

Packaging

Foil pouches

Reconstitution

Contents of one pouch, when dissolved in 500 mL of ultrapure water, will yield a 0.1M Tris solution, pH 8.1 at 25 °C. Contents tested to be DNAse, RNAse, Protease, and Nickase free. This product is suitable for use in cell culture and electrophoresis applications.

Storage and Stability

Store at room temperature. Product may naturally agglomerate but can be simply broken up within the pouch prior to use.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

pHast Pack is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service