Skip to Content
Merck
All Photos(1)

Key Documents

PHR8952

Supelco

Mirtazapine

new

certified reference material, pharmaceutical secondary standard

Synonym(s):

Mirtazapine, 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3
CAS Number:
Molecular Weight:
265.35
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Agency

traceable to Ph. Eur. Y0000705
traceable to USP 1444279

API family

mirtazapine

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

CN1CCN2C(C1)c3ccccc3Cc4cccnc24

InChI

1S/C17H19N3/c1-19-9-10-20-16(12-19)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)20/h2-8,16H,9-12H2,1H3

InChI key

RONZAEMNMFQXRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Biochem/physiol Actions

Mirtazapine is a noradrenergic and specific serotonergic antidepressant (NaSSA). Mirtazapine agonizes selective adrenergic and serotonergic receptors so that both NE release and 5-HT1A mediated serotonergic signaling are increased.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service