Skip to Content
Merck
All Photos(2)

Key Documents

PHR8631

Supelco

Quetiapine Related Compound H

certified reference material, pharmaceutical secondary standard

Synonym(s):

Quetiapine N-oxide, 2-[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-oxido-1-piperazinyl)ethoxy]ethanol, 4-(Dibenzo[b,f][1,4]thiazepin-11-yl)-1-[2-(2-hydroxyethoxy)ethyl]piperazine 1-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N3O3S
CAS Number:
Molecular Weight:
399.51
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available. Contact Technical Service

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1597292

API family

quetiapine

packaging

pkg of 50MG

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

[s]1c2c(nc(c4c1cccc4)N3CC[N+](CC3)([O-])CCOCCO)cccc2

InChI

1S/C21H25N3O3S/c25-14-16-27-15-13-24(26)11-9-23(10-12-24)21-17-5-1-3-7-19(17)28-20-8-4-2-6-18(20)22-21/h1-8,25H,9-16H2

InChI key

VVBXDYFGQNNTDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service