Skip to Content
Merck
All Photos(2)

Key Documents

PHR3857

Supelco

(S)-β-Amino-3-[2-(dimethylamino)ethyl]-1H-indole-5-propanol

certified reference material, pharmaceutical secondary standard

Synonym(s):

(S)-2-Amino-3-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}propan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N3O
CAS Number:
Molecular Weight:
261.36
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1727031

API family

zolmitriptan

packaging

pkg of 50MG

application(s)

pharmaceutical

SMILES string

[nH]1c2c(c(c1)CCN(C)C)cc(cc2)C[C@H](N)CO

InChI

1S/C15H23N3O/c1-18(2)6-5-12-9-17-15-4-3-11(8-14(12)15)7-13(16)10-19/h3-4,8-9,13,17,19H,5-7,10,16H2,1-2H3/t13-/m0/s1

InChI key

XYSYRJCYRKMOFP-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service