Skip to Content
Merck
All Photos(1)

Key Documents

PHR3650

5′ -Cytidylic Acid

new

certified reference material, pharmaceutical secondary standard

Synonym(s):

Cytidine 5′-monophosphate, 5′-CMP, 5′-Cytidylic acid, C-5′-P

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N3O8P
CAS Number:
Molecular Weight:
323.20
Beilstein:
46982
MDL number:

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1162126

packaging

pkg of 1G

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]2O

InChI

1S/C9H14N3O8P/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18/h1-2,4,6-8,13-14H,3H2,(H2,10,11,15)(H2,16,17,18)/t4-,6-,7-,8-/m1/s1

InChI key

IERHLVCPSMICTF-XVFCMESISA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service