Skip to Content
Merck
All Photos(1)

Key Documents

PHR3543

Supelco

Nonanal

certified reference material, pharmaceutical secondary standard

Synonym(s):

Aldehyde C9, Nonyl aldehyde, Pelargonaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CHO
CAS Number:
Molecular Weight:
142.24
Beilstein:
1236701
MDL number:
UNSPSC Code:
41116107

grade

certified reference material
pharmaceutical secondary standard

vapor pressure

~0.26 mmHg ( 25 °C)

packaging

pkg of 1 g

refractive index

n20/D 1.424 (lit.)

bp

93 °C/23 mmHg (lit.)

density

0.827 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCCCCCC=O

InChI

1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h9H,2-8H2,1H3

InChI key

GYHFUZHODSMOHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6959(changes by product) in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service