Skip to Content
Merck
All Photos(1)

Key Documents

PHR2731

Supelco

Nonoxynol 9

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

α-(4-Nonylphenyl)-ω-hydroxynona(oxyethylene), 2-(4-Nonylphenoxy)ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€143.00

€143.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€143.00

About This Item

Linear Formula:
C17H28O2
UNSPSC Code:
41116107
NACRES:
NA.24

€143.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000755

API family

nonoxynol

form

liquid

packaging

pkg of 1 g

application(s)

pharmaceutical small molecule

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6959 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hi morning, Recently we have bought Nonoxynol 9 (PHR2731-1G) but is liquid. I would like to know what the concentration is. Thanks in advance.

    1 answer
    1. This product is a neat liquid with a purity of 98.8%. The average molecular weight is 616.82 g/mol, and the density is 0.960 g/mL. The molarity is calculated as (960 g/L ÷ 616.82 g/mol) * 0.9888 = 1.54 M.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service