PHR2148
Phenylephrine Related Compound D
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(R-3-{2-[Benzyl(methyl)amino]-1-hydroxyethyl}phenol
Select a Size
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1533035
API family
phenylephrine
CofA
current certificate can be downloaded
packaging
pkg of 100 mg
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
N(C[C@H](O)c2cc(ccc2)O)(Cc1ccccc1)C
InChI
1S/C16H19NO2/c1-17(11-13-6-3-2-4-7-13)12-16(19)14-8-5-9-15(18)10-14/h2-10,16,18-19H,11-12H2,1H3/t16-/m0/s1
InChI key
JBBZPOHBHNEKIM-INIZCTEOSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Recommended products
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service