Skip to Content
Merck

Skip To

PHR2002

Folic Acid Impurity D

Pharmaceutical Secondary Standard; Certified Reference Material

Promotion Available

Synonym(s):

Pteroic acid

Sign In to View Organizational & Contract Pricing.

Spend € 450 — Save 5 %; Spend € 700 — Save 10 %; Spend € 1.500 — Save 15 %*

Sign in to check eligibility. Shop Products

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 704,00

About This Item

Empirical Formula (Hill Notation):
C14H12N6O3
CAS Number:
Molecular Weight:
312.28
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

€ 704,00

Save Up To 15 %


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, pharmaceutical secondary standard

agency

traceable to Ph. Eur. Y0001243

API family

folic acid

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

NC1=NC(=O)C2=NC(CNc3ccc(cc3)C(O)=O)=CNC2=N1

InChI

1S/C14H12N6O3/c15-14-19-11-10(12(21)20-14)18-9(6-17-11)5-16-8-3-1-7(2-4-8)13(22)23/h1-4,6,16H,5H2,(H,22,23)(H3,15,17,19,20,21)

InChI key

JOAQINSXLLMRCV-UHFFFAOYSA-N

General description

Folic Acid Impurity D is an impurity of the water-soluble B-vitamin, folic acid. Folic Acid is generally used in the treatment of certain types of anemia.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Folic Acid Impurity D may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by high performance liquid chromatography.

Analysis Note

These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
To see an example of a Certificate of Analysis for this material enter LRAB4757 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P1781PHR2003PHR1334
description

Pharmaceutical Secondary Standard; Certified Reference Material

description

≥93%

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

Pharmaceutical Secondary Standard; Certified Reference Material

application(s)

pharmaceutical

application(s)

-

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

-

format

neat

Quality Level

300

Quality Level

200

Quality Level

300

Quality Level

300

grade

pharmaceutical secondary standard, certified reference material

grade

-

grade

pharmaceutical secondary standard, certified reference material

grade

certified reference material, pharmaceutical secondary standard

storage temp.

2-8°C

storage temp.

-

storage temp.

2-30°C

storage temp.

2-30°C

packaging

pkg of 100 mg

packaging

-

packaging

pkg of 20 mg

packaging

-


Still not finding the right product?

Explore all of our products under Folic Acid Impurity D


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters