Skip to Content
Merck
All Photos(1)

Key Documents

PHR1840

Supelco

Aminobenzoic Acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4-Aminobenzoic acid, PABA, Vitamin Bx, Vitamin H1

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€170.00

€170.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€170.00

About This Item

Linear Formula:
H2NC6H4CO2H
CAS Number:
Molecular Weight:
137.14
Beilstein:
471605
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€170.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000173
traceable to USP 1019803

API family

aminobenzoic acid

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

mp

187-189 °C (lit.)

density

1.374 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccc(cc1)C(O)=O

InChI

1S/C7H7NO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H,9,10)

InChI key

ALYNCZNDIQEVRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Amiodarone is an antiarrhythmic agent used for the prophylaxis and treatment of ventricular arrhythmias and atrial fibrillation.[1]

Application

Aminobenzoic Acid may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical preparation by high performance liquid chromatographic method.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2774 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

339.8 °F - closed cup

Flash Point(C)

171 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of nicotinamide and 4-aminobenzoic acid in pharmaceutical preparation by LC
Khan AR, et al.
Journal of Pharmaceutical and Biomedical Analysis, 29(4), 723-727 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service