Skip to Content
Merck
All Photos(1)

Key Documents

PHR1828

Supelco

Eucalyptol (cineole)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Eucalyptol, 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane, 1,8-Cineole, 1,8-Epoxy-p-menthane

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€205.00

€205.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
105109
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.24

€205.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000176
traceable to USP 1268900

API family

cineole

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.457 (lit.)

bp

176-177 °C (lit.)

mp

1-2 °C (lit.)

density

0.921 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H](CC1)C(C)(C)O2

InChI

1S/C10H18O/c1-9(2)8-4-6-10(3,11-9)7-5-8/h8H,4-7H2,1-3H3/t8-,10+

InChI key

WEEGYLXZBRQIMU-WAAGHKOSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Eucalyptol, a natural monoterpene, is a component of many medicinally active aromatic plants, such as eucalyptus, cardamom, thyme, rosemary, and ginger. It exhibits subduing effect on mucus hypersecretion, non-purulent rhinosinusitis, and asthma.[1]

Application

This pharmaceutical secondary standard can also be used as follows:

  • Simultaneous estimation of menthol and eucalyptol in their combined marketed analgesic formulations by high-performance thin layer chromatography (HPTLC) procedure[1]
  • The development of an International Conference on Harmonization (ICH) guideline validated HPTLC method for the determination of α-pinene and eucalyptol in plants from the Lamiaceae family[2]
  • Headspace sorptive extraction (HSSE) of seven monoterpenes for their gas chromatography and mass spectrometry (GC-MS) based quantitative analysis from different honey samples[3]
  • Screening and estimation of eucalyptol in a polyherbal formulation using an HPTLC-based technique[4]

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB9507 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of high-performance thin-layer chromatography (HPTLC) validated method for simultaneous quantification of eucalyptol and ?-pinene in lamiaceae plants
Foudah AI, et al.
Journal of Pharmaceutical Research International, 31, 1-11 (2019)
A validated HPTLC method for estimation of Eucalyptol in polyherbal formulations
Patra KC, et al.
Der Pharmacia Lettre, 2, 316-320 (2010)
J I Cacho et al.
Journal of chromatography. A, 1399, 18-24 (2015-05-11)
Headspace sorptive extraction (HSSE) was used to preconcentrate seven monoterpenes (eucalyptol, linalool, menthol, geraniol, carvacrol, thymol and eugenol) for separation by gas chromatography and mass spectrometry (GC-MS). Three commercially available coatings for the stir bars, namely Polydimethylsiloxane (PDMS), polyacrilate (PA)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service