Skip to Content
Merck
All Photos(1)

Key Documents

PHR1756

Supelco

Bupivacaine Related Compound B

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Desbutylbupivacaine, (2RS)-N-(2,6-dimethylphenyl)piperidine-2-carboxamide, (RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
€692.00

€692.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
€692.00

About This Item

Empirical Formula (Hill Notation):
C14H20N2O
CAS Number:
Molecular Weight:
232.32
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€692.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000088
traceable to USP 1078529

API family

bupivacaine

CofA

current certificate can be downloaded

packaging

pkg of 20 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(NC1=C(C)C=CC=C1C)C2NCCCC2

InChI

1S/C14H20N2O/c1-10-6-5-7-11(2)13(10)16-14(17)12-8-3-4-9-15-12/h5-7,12,15H,3-4,8-9H2,1-2H3,(H,16,17)

InChI key

SILRCGDPZGQJOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Bupivacaine Related Compound B is formed in the liver during oxidative dealkylation of bupivacaine hydrochloride and is the main metabolite of bupivacaine. It is a local anesthetic agent that can cross biological membranes due to its lipid-soluble nature.[1]

Application

Bupivacaine may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatographic[2] and spectrophotometric[3] techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7918 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of bupivacaine and its two metabolites, desbutyl-and 4′-hydroxybupivacaine, in human serum and urine
Lindberg RLP, et al
Journal of Chromatography. B, Biomedical Sciences and Applications, 383, 357-364 (1986)
Determination of tramadol, metamizole, ropivacaine, and bupivacaine in analgesic mixture samples by HPLC with DAD detection
Salmeron-Garcia A, et al.
Journal of Chromatographic Science, 47(3), 231-237 (2009)
Quantitative mass spectrometric analysis of ropivacaine and bupivacaine in authentic, pharmaceutical and spiked human plasma without chromatographic separation
Salama NN and Wang S
Analytical Chemistry Insights, 4(3), ACI-S2564 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service