Skip to Content
Merck
All Photos(1)

Key Documents

PHR1668

Supelco

Retinyl Propionate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Retinyl propionate, Vitamin A propionate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€126.00

€126.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€126.00

About This Item

Empirical Formula (Hill Notation):
C23H34O2
CAS Number:
Molecular Weight:
342.51
Beilstein:
2475675
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€126.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

retinyl compounds

form

liquid

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC(=O)OC\C=C(C)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C23H34O2/c1-7-22(24)25-17-15-19(3)11-8-10-18(2)13-14-21-20(4)12-9-16-23(21,5)6/h8,10-11,13-15H,7,9,12,16-17H2,1-6H3/b11-8+,14-13+,18-10+,19-15+

InChI key

SFRPDSKECHTFQA-ONOWFSFQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards. Retinyl Propionate belongs to the group of retinyl esters[1] and is found to be active in human skin.[2] It shows cosmetic related efficacy and hence helps in the improvement of photodamaged facial skin.[2]

Application

Retinyl Propionate may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using liquid chromatography technique.[3]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB5021 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Cosmetic Medicine and Surgery, An Issue of Clinics in Plastic Surgery - E- Book (2011)
An isocratic LC method for the simultaneous determination of vitamins A, C, E and ?-carotene
Paulo GM, et al.
Journal of Pharmaceutical and Biomedical Analysis, 21(2), 399-406 (1999)
Cosmeceuticals E-Book: Procedures in Cosmetic Dermatology Series, 21(3), 399-406 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service