Skip to Content
Merck
All Photos(1)

Key Documents

PHR1658

Supelco

Dimenhydrinate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Dimenhydrinate, DMH, N-(2-Diphenylmethoxyethyl)-N,N-dimethylammonium 8-chlorotheophyllinate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€129.00

€129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€129.00

About This Item

Empirical Formula (Hill Notation):
C17H21NO · C7H7ClN4O2
CAS Number:
Molecular Weight:
469.96
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€129.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. D2000000
traceable to USP 1206006

API family

dimenhydrinate

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CN1C(=O)N(C)c2nc(Cl)[nH]c2C1=O.CN(C)CCOC(c3ccccc3)c4ccccc4

InChI

1S/C17H21NO.C7H7ClN4O2/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;1-11-4-3(9-6(8)10-4)5(13)12(2)7(11)14/h3-12,17H,13-14H2,1-2H3;1-2H3,(H,9,10)

InChI key

NFLLKCVHYJRNRH-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Dimenhydrinate, a diphenhydramine salt of 8-chlorotheophylline, exhibits antihistaminic activity, with antimuscarinic and sedative effects. It finds its major application as an antiemetic drug in the prevention and treatment of motion sickness.[1]

Application

Dimenhydrinate may be used as a pharmaceutical reference standard for the determination of the analyte in traditional Chinese medicine[2] and pharmaceutical formulations[3][1] by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3017 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of Dimenhydrinate, Cinnarizine and Cinnarizine impurity by TLC and HPLC chromatographic methods.
Ahmed AB, et al.
Bulletin of Faculty of Pharmacy, Cairo University , 55(1), 163-169 (2017)
Optimization and validation of a method for the determination of caffeine, 8-chlorotheophylline and diphenhydramine by isocratic high-performance liquid chromatography: Stress test for stability evaluation.
Barbas C, et al.
Journal of Chromatography A, 870(1-2), 97-103 (2000)
HPLC and GC?MS screening of Chinese proprietary medicine for undeclared therapeutic substances.
Liu SY, et al.
Journal of Pharmaceutical and Biomedical Analysis, 24(5-6), 983-992 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service