Skip to Content
Merck
All Photos(1)

Key Documents

PHR1591

Supelco

Atovaquone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Atovaquone, Mepron, trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthalenedione

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€107.00

€107.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
500 MG
€107.00

About This Item

Empirical Formula (Hill Notation):
C22H19ClO3
CAS Number:
Molecular Weight:
366.84
UNSPSC Code:
41116107
NACRES:
NA.24

€107.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001572
traceable to USP 1044651

API family

atovaquone

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC1=C([C@H]2CC[C@@H](CC2)c3ccc(Cl)cc3)C(=O)c4ccccc4C1=O

InChI

1S/C22H19ClO3/c23-16-11-9-14(10-12-16)13-5-7-15(8-6-13)19-20(24)17-3-1-2-4-18(17)21(25)22(19)26/h1-4,9-13,15,26H,5-8H2/t13-,15-

InChI key

KUCQYCKVKVOKAY-CTYIDZIISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Atovaquone belongs to the class of naphthoquinones with a broad-spectrum activity against parasitic infections including malaria, toxoplasmosis and Pneumocystis pneumonia. It is a potent antiprotozoal compound which aids in collapsing the mitochondrial membrane potential in a malaria parasite.[1]

Application

Atovaquone may be used as a pharmaceutical secondary reference standard for the determination of the analyte in plasma samples by liquid-liquid extraction followed by reversed-phase high-performance liquid chromatography with ultraviolet detection.[2]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Atovaquone in an anti-protozoal mitochondrial electron transport inhibitor. It also functions as an antimalarial and antipneumocystic agent.
Atovaquone is an anti-protozoal mitochondrial electron transport inhibitor; Antimalarial; Antipneumocystic, and has also been used to treat toxoplasmosis. It is an analog of protozoan mitochondrial protein ubiquinone, and acts by inhibiting the cytochrome bc(1) complex via interactions with the Rieske iron-sulfur protein and cytochrome b in the ubiquinol oxidation pocket.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA4707 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of the potent antiprotozoal compound atovaquone in plasma using liquid-liquid extraction followed by reversed-phase high-performance liquid chromatography with ultraviolet detection
Hannan SL, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 678(2), 297-302 (1996)
Antiparasitic agent atovaquone
Baggish AL and Hill DR
Antimicrobial Agents and Chemotherapy, 46(5), 1163-1173 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service