Skip to Content
Merck
All Photos(1)

Documents

PHR1496

Supelco

Paromomycin Sulfate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Paromomycin sulfate salt, Aminosidine sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H45N5O14 · xH2SO4
CAS Number:
Molecular Weight:
615.63 (free base basis)
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1500003

API family

paromomycin

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

InChI

1S/C23H45N5O14.H2O4S/c24-2-7-13(32)15(34)10(27)21(37-7)41-19-9(4-30)39-23(17(19)36)42-20-12(31)5(25)1-6(26)18(20)40-22-11(28)16(35)14(33)8(3-29)38-22;1-5(2,3)4/h5-23,29-36H,1-4,24-28H2;(H2,1,2,3,4)/t5-,6+,7+,8-,9-,10-,11-,12+,13-,14-,15-,16-,17-,18?,19-,20-,21-,22-,23?;/m1./s1

InChI key

LJRDOKAZOAKLDU-UMIPPVEZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Paromomycin Sulfate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by potentiometric as well as high performance liquid chromatography (HPLC) techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3044 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of salbutamol, amikacin and paromomycin sulfate by a novel sulfate polymeric membrane sensor based on 2, 6-diphenyl 4-(4-methoxyphenyl) pyrylium perchlorate.
Ganjali MR, et al.
Microchimica Acta, 149(3-4), 245-249 (2005)
Development a new method for the determination of paromomycin in trace amounts by fast Fourier continuous cyclic voltammetry at an Au microelectrode in a flowing system.
Ganjali MR, et al.
Sensors and Actuators B, Chemical, 123(2), 1125-1132 (2007)
Toshiyuki Oda et al.
Molecular biology of the cell, 26(2), 294-304 (2014-11-21)
The nexin-dynein regulatory complex (N-DRC) forms a cross-bridge between the outer doublet microtubules of the axoneme and regulates dynein motor activity in cilia/flagella. Although the molecular composition and the three-dimensional structure of N-DRC have been studied using mutant strains lacking
Alexandre Melnikov et al.
Nucleic acids research, 42(14), e112-e112 (2014-06-11)
Deep mutational scanning has emerged as a promising tool for mapping sequence-activity relationships in proteins, ribonucleic acid and deoxyribonucleic acid. In this approach, diverse variants of a sequence of interest are first ranked according to their activities in a relevant
Walid M Fouad et al.
Current pharmaceutical biotechnology, 16(5), 407-413 (2015-03-10)
Lignocellulosic biomass has the potential to serve as feedstock and direct replacement for petrochemicals in the fuel, chemical, pharmaceutical and material industries. Energy cane has been identified by the U.S. Department of Energy (DOE) as prime lignocellulosic feedstock as it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service