Skip to Content
Merck
All Photos(1)

Key Documents

PHR1297

Supelco

Phosphated Riboflavin (Riboflavin Sodium Phosphate)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Riboflavin 5′-monophosphate sodium salt hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€93.10

€93.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€93.10

About This Item

Empirical Formula (Hill Notation):
C17H20N4O9P · xNa+ · yH2O
Molecular Weight:
455.34 (anhydrous free acid basis)
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€93.10


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 459
traceable to Ph. Eur. R0630000
traceable to USP 1535700

API family

riboflavin

CofA

current certificate can be downloaded

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Phosphated Riboflavin (Riboflavin Sodium Phosphate) may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.[1][2]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0584 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The simultaneous assay of riboflavin 5-phosphate sodium and other water-soluble vitamins in liquid multivitamin formulations by liquid chromatograph
Lam FL and Lowande A
Journal of Pharmaceutical and Biomedical Analysis, 6(1), 87-95 (1988)
Simultaneous determination of riboflavin phosphate and other ingredients in a multivitamin pharmaceutical preparation by on-line automated LC coupled with pre-column immobilized enzyme reactor
Ono M, et al.
Journal of Pharmaceutical and Biomedical Analysis, 29(1-2), 325-334 (2002)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service