Skip to Content
Merck
All Photos(1)

Key Documents

PHR1250

Supelco

Povidone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Polyvinylpyrrolidone, PVP, Polyvidone, Povidone

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€95.80

€95.80


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
500 MG
€95.80

About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€95.80


Estimated to ship on14 April 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. P2660000
traceable to USP 1551503

API family

povidone

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Povidone may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.[1][2][3][4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3638 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultra rapid liquid chromatography as second dimension in a comprehensive two-dimensional method for the screening of pharmaceutical samples in stability and stress studies
Huidobro AL, et al.
Journal of Chromatography A, 1190(1-2), 182-190 (2008)
Determination of bile acids in pharmaceutical formulations using micellar electrokinetic chromatography
Rodriguez VG, et al.
Journal of Pharmaceutical and Biomedical Analysis, 23(2-3), 375-381 (2000)
The simultaneous separation and determination of five quinolone antibotics using isocratic reversed-phase HPLC: Application to stability studies on an ofloxacin tablet formulation
Shervington LA, et al.
Journal of Pharmaceutical and Biomedical Analysis, 39(3-4), 769-775 (2005)
Detection and quantification of low-molecular-weight aldehydes in pharmaceutical excipients by headspace gas chromatography
Li Z, et al.
Journal of Chromatography A, 1104(1-2), 1-10 (2006)
X C Qin et al.
Journal of photochemistry and photobiology. B, Biology, 120, 156-162 (2013-01-30)
Nanographene oxide (NGO), a new type of nanomaterial for anticancer drugs delivery and near-infrared (NIR)-mediated photothermal ablation of tumors, has been used in the combination of photothermal therapy and chemotherapy. Herein, targeted chemo-photothermal therapy based on polyvinylpyrrolidone (PVP) functionalized NGO

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service