Skip to Content
Merck
All Photos(1)

Key Documents

PHR1146

Supelco

Ibuprofen Related Compound C

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4′-Isobutylacetophenone, 1-(4-Isobutylphenyl)ethanone, 1-Acetyl-4-isobutylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€111.00

€111.00


Estimated to ship on06 April 2025


Request a Bulk Order

Select a Size

Change View
500 MG
€111.00

About This Item

Empirical Formula (Hill Notation):
C12H16O
CAS Number:
Molecular Weight:
176.25
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€111.00


Estimated to ship on06 April 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

100
300

Agency

traceable to BP 557
traceable to USP 1335541

API family

ibuprofen

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(C)c1ccc(cc1)CC(C)C

InChI

1S/C12H16O/c1-9(2)8-11-4-6-12(7-5-11)10(3)13/h4-7,9H,8H2,1-3H3

InChI key

KEAGRYYGYWZVPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

It is an impurity of the nonsteroidal anti-inflammatory drug (NSAID)― ibuprofen and presents itself as a degradation product of the parent drug. It is used for the treatment of mild and moderate pain such as during rheumatoid arthritis, osteoarthritis, and dysmenorrhea.[1]

Application

This pharmaceutical secondary standard can also be used as follows:

  • Simultaneous determination of ibuprofen and its 17 related impurities by an ICH validated reversed-phase high-performance liquid chromatography (RP-HPLC) method in tablets[1]
  • Analysis of combined dosage tablet of ibuprofen and famotidine for their quantitative estimation in the presence of their impurities using thin layer chromatography (TLC) and HPLC

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB2151 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

286.7 °F - closed cup

Flash Point(C)

141.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of an HPLC method for simultaneous determination of ibuprofen and 17 related compounds
Han Z, et al.
Chromatographia, 80, 1353-1360 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service