Skip to Content
Merck
All Photos(1)

Key Documents

PHR1123

Supelco

Isopropyl myristate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Isopropyl tetradecanoate, Myristic acid isopropyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€122.00

€122.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€122.00

About This Item

Linear Formula:
CH3(CH2)12COOCH(CH3)2
CAS Number:
Molecular Weight:
270.45
Beilstein:
1781127
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€122.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. I0750000
traceable to USP 1350400

API family

isopropyl myristate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.434 (lit.)

bp

193 °C/20 mmHg (lit.)

mp

~3 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCC(=O)OC(C)C

InChI

1S/C17H34O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-17(18)19-16(2)3/h16H,4-15H2,1-3H3

InChI key

AXISYYRBXTVTFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropyl myristate is an ester of isopropyl alcohol myristic acid.[1] It is mainly used as a solubilizer, emulsifier and emollient in cosmetic and topical medicines. It also finds applications as a flavoring agent in the food industry.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7001 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

>302.0 °F

Flash Point(C)

> 150 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CRC Handbook of Food, Drug, and Cosmetic Excipients (1992)
T N Engelbrecht et al.
Skin pharmacology and physiology, 25(4), 200-207 (2012-05-23)
In order to elucidate the mode of action of the lipophilic penetration enhancer isopropyl myristate (IPM) on a molecular scale, we investigated oriented quaternary stratum corneum (SC) lipid model membranes based on ceramide AP, cholesterol, palmitic acid and cholesterol sulfate
Qingping Tian et al.
International journal of pharmaceutics, 426(1-2), 202-210 (2012-01-26)
To improve the skin permeation of naproxen with larger dosage, microemulsion with high content of naproxen was investigated for transdermal delivery and its solubilization mechanism was studied. Naproxen micoremulsions composed of 4% isopropyl myristate, 18% Tween 80, 18% ethanol and
Fan Yang et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 82(1), 158-163 (2012-06-21)
Transdermal delivery of methotrexate (MTX) was investigated by using the solid-in-oil (S/O) technique. Because MTX was coated with nonionic surfactant molecules, the resulting complex was easy to dissolve in various organic solvents and provided a transparent solution in isopropyl myristate
Qian Zhang et al.
Pharmaceutical research, 30(1), 32-40 (2012-08-28)
In principle, maximum transepidermal fluxes of solutes should be similar for different vehicles, except when the solute or vehicle modifies the skin. Here we estimated maximum flux, stratum corneum solubility, diffusivity and permeability coefficient for a range of similarly sized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service