Skip to Content
Merck
All Photos(1)

Key Documents

PHL89156

Alantolactone

phyproof® Reference Substance

Synonym(s):

(+)-Alantolactone., 3aR,5S,8aR,9aR)-3a,5,6,7,8,8a,9,9a-Octahydro-5,8a-dimethyl-3-methylene-naphtho[2,3-b]furan-2(3H)-one, Alant camphor, Elecampane camphor, Eupatal, Inula camphor, NSC 333843, NSC 93131

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€379.00

€379.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€379.00

About This Item

Empirical Formula (Hill Notation):
C15H20O2
CAS Number:
Molecular Weight:
232.32
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€379.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

C[C@H]1CCC[C@]2(C)C[C@H]3OC(=O)C(=C)[C@H]3C=C12

InChI

1S/C15H20O2/c1-9-5-4-6-15(3)8-13-11(7-12(9)15)10(2)14(16)17-13/h7,9,11,13H,2,4-6,8H2,1,3H3/t9-,11+,13+,15+/m0/s1

InChI key

PXOYOCNNSUAQNS-AGNJHWRGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Alantolactone is a sesquiterpene lactone disrupts the Cripto-1/ActRII complexes, resulting in an induction of activin/SMAD3 signaling. Alantolactone inhibits proliferation in cancer cells with no affect on normal cells.
Alantolactone is an inducer of activin/SMAD3 signaling.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaomin Dang et al.
Respiratory research, 21(1), 95-95 (2020-04-24)
It is well established that airway remodeling and inflammation are characteristics for chronic obstructive pulmonary disease (COPD). Moreover, cigarette smoke extract (CSE) promots inflammation, apoptosis and oxidative stress in COPD. And, there is evidence suggested that alantolactone (ALT), a sesquiterpene
Hsiang-Lai Chen et al.
Pharmaceutical biology, 59(1), 134-145 (2021-02-09)
Alantolactone, the bioactive component in Inula helenium L. (Asteraceae), exhibits multiple biological effects. We aimed to determine the anti-inflammatory effect of alantolactone in a collagen-induced arthritis (CIA) mouse model and its immunomodulatory effects on Th17 differentiation. A CIA mouse model

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service