Skip to Content
Merck
All Photos(1)

Key Documents

PHL80478

Icaritin

phyproof® Reference Substance

Synonym(s):

3,5,7-Trihydroxy-2-(4-methoxyphenyl)-8-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€408.00

€408.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€408.00

About This Item

Empirical Formula (Hill Notation):
C21H20O6
CAS Number:
Molecular Weight:
368.38
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€408.00


Estimated to ship on24 May 2025


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C2=C(O)C(=O)c3c(O)cc(O)c(C\C=C(\C)C)c3O2

InChI

1S/C21H20O6/c1-11(2)4-9-14-15(22)10-16(23)17-18(24)19(25)20(27-21(14)17)12-5-7-13(26-3)8-6-12/h4-8,10,22-23,25H,9H2,1-3H3

InChI key

TUUXBSASAQJECY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Icaritin enhances osteoblastic differentiation of mesenchymal stem cells (MSCs) while it inhibits adipogenic differentiation of MSCs.
Icaritin is a component of Epimedium flavonoid isolated from Herba Epimedii, which enhances osteoblastic differentiation of mesenchymal stem cells (MSCs) while it inhibits adipogenic differentiation of MSCs by inhibiting PPAR-g pathway. Icaritin has no effect on MSCs proliferation. Also, icaritin potently inhibits chronic myeloid leukemia (CML) and breast cancer cells proliferation most likely by modulation of MAPK/ERK/JNK and JAK2/STAT3 /AKT signaling. As other flavonoids, icaritin may exert estrogen-like activities.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service