Skip to Content
Merck

Skip To

PHL80452

Fangchinoline

phyproof® Reference Substance

Synonym(s):

(+)-Limacine, 7-O-Demethyltetrandine, Frangchinoline, Limacine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 199,00

About This Item

Empirical Formula (Hill Notation):
C37H40N2O6
CAS Number:
Molecular Weight:
608.72
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

€ 199,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

SMILES string

CN([C@@]1([H])CC(C=C2)=CC=C2OC3=C(OC)C=CC4=C3)CCC5=C1C=C(OC6=C([C@@](C4)([H])N(C)CC7)C7=CC(OC)=C6O)C(OC)=C5

InChI

1S/C37H40N2O6/c1-38-14-12-24-19-31(42-4)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-32-18-23(8-11-30(32)41-3)17-29-35-25(13-15-39(29)2)20-34(43-5)36(40)37(35)45-33/h6-11,18-21,28-29,40H,12-17H2,1-5H3/t28-,29-/m0/s1

InChI key

IIQSJHUEZBTSAT-VMPREFPWSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHL83302PHL89273PHL85707
grade

primary reference standard

grade

primary reference standard

grade

primary reference standard

grade

primary reference standard

format

neat

format

neat

format

neat

format

neat

assay

≥95.0% (HPLC)

assay

≥95.0% (HPLC)

assay

≥98.0% (HPLC)

assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

manufacturer/tradename

PhytoLab

manufacturer/tradename

PhytoLab

manufacturer/tradename

PhytoLab

product line

phyproof® Reference Substance

product line

phyproof® Reference Substance

product line

phyproof® Reference Substance

product line

phyproof® Reference Substance


Still not finding the right product?

Explore all of our products under Fangchinoline


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters