Skip to Content
Merck
All Photos(1)

Key Documents

PHL80440

Ergocristine

phyproof® Reference Substance

Synonym(s):

12′-Hydroxy-2′-(1-methylethyl)-5′-(phenylmethyl)ergotaman-3′,6′,18′-trione

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€449.00

€449.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€449.00

About This Item

Empirical Formula (Hill Notation):
C35H39N5O5
CAS Number:
Molecular Weight:
609.71
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€449.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

mp

155-157 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

CC(C)[C@@]5(NC(=O)[C@H]1CN(C)[C@@H]2Cc3c[nH]c4cccc(C2=C1)c34)O[C@@]6(O)[C@@H]7CCCN7C(=O)[C@H](Cc8ccccc8)N6C5=O

InChI

1S/C35H39N5O5/c1-20(2)34(37-31(41)23-16-25-24-11-7-12-26-30(24)22(18-36-26)17-27(25)38(3)19-23)33(43)40-28(15-21-9-5-4-6-10-21)32(42)39-14-8-13-29(39)35(40,44)45-34/h4-7,9-12,16,18,20,23,27-29,36,44H,8,13-15,17,19H2,1-3H3,(H,37,41)/t23-,27-,28+,29+,34-,35+/m1/s1

InChI key

HEFIYUQVAZFDEE-MKTPKCENSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Navneet Kaur et al.
Journal of economic entomology, 113(5), 2079-2085 (2020-07-12)
Our previous study provided correlative evidence that morning glory species harboring endophytic fungi (Periglandula) are resistant to potato psyllid [Bactericera cockerelli (Šulc)], whereas species free of fungi often allowed psyllid development. In this study, we manipulated levels of ergot alkaloids
Céline Peillod et al.
Toxins, 13(2) (2021-03-07)
Toxic effects among fumonisins B (FB), deoxynivalenol (DON) and zearalenone (ZEN) administered alone and combined were investigated in 84-day-old ducks during force-feeding. 75 male ducks, divided into five groups of 15 animals, received daily during the meal a capsule containing
D Tardieu et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 148, 111968-111968 (2021-01-11)
Previous studies using zearalenone (ZEN) and fumonisins (FB) revealed alpha-zearalanol (α-ZOL) and FB1 in the liver of turkeys and chickens with no sign of toxicity. The aim of the present study was to determine whether contamination persists after distribution of
Angélique Travel et al.
Avian diseases, 63(4), 703-712 (2019-12-24)
Surveys of mycotoxins worldwide have shown that deoxynivalenol (DON), fumonisins (FB), and zearalenone (ZON) are the most abundant Fusarium mycotoxins (FUS) in European poultry feed, in both the level and the frequency of contamination. Previous studies reported that a combination

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service