Skip to Content
Merck
All Photos(1)

Key Documents

PHL80181

Urushiol (15:2)

phyproof® Reference Substance

Synonym(s):

3-(8Z,11Z-Pentadecadienyl)-1,2-benzenediol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€593.00

€593.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€593.00

About This Item

Empirical Formula (Hill Notation):
C21H32O2
CAS Number:
Molecular Weight:
316.48
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€593.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥85.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

Oc1c(cccc1CCCCCCC\C=C/C\C=C/CCC)O

InChI

1S/C21H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-17-15-18-20(22)21(19)23/h4-5,7-8,15,17-18,22-23H,2-3,6,9-14,16H2,1H3/b5-4-,8-7-

InChI key

RMTXUPIIESNLPW-UTOQUPLUSA-N

Related Categories

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Does PHL80181 contain stabilizer? What techniques is it suitable for?

    1 answer
    1. The PHL80181 Urushiol (15:2) does not contain stabilizers and is suitable for use in LC-MS, GC-MS, and ESI-MS applications.

      Helpful?

  2. Does product PHL80181 Urushiol (15:2) contain stabilizer? What applications is it suitable for?

    1 answer
    1. PHL80181 Urushiol (15:2) does not contain stabilizers and is suitable for LC-MS, GC-MS, and ESI-MS applications.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service