Skip to Content
Merck
All Photos(3)

Key Documents

P6474

Sigma-Aldrich

TWEEN® 80

viscous liquid, Preservative Free, Low-peroxide; Low-carbonyls

Synonym(s):

Tween® 80KR, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€152.00
5 X 10 ML
€548.00
100 ML
€881.00

€152.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€152.00
5 X 10 ML
€548.00
100 ML
€881.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

€152.00


Check Cart for Availability

Request a Bulk Order

Quality Level

description

non-ionic

form

viscous liquid

mol wt

micellar avg mol wt 79,000
average mol wt 1310

composition

Oleic acid, ~70% (balance primarily linoleic, palmitic, and stearic acids)

aggregation number

60

impurities

≤0.5 μmol/g Peroxides
≤1.0 μmol/g Carbonyls
≤3.0% water

CMC

0.012 mM (20-25°C)

transition temp

cloud point 65 °C

density

1.06 g/mL at 25 °C

HLB

15

storage temp.

−20°C

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-ionic detergent used for selective protein extraction and isolation of nuclei from mammalian cell lines.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie J Alexopoulos et al.
Nature communications, 11(1), 2397-2397 (2020-05-16)
Obesity is a health problem affecting more than 40% of US adults and 13% of the global population. Anti-obesity treatments including diet, exercise, surgery and pharmacotherapies have so far failed to reverse obesity incidence. Herein, we target obesity with a
Qun Wang et al.
Brain research, 1285, 188-195 (2009-06-17)
The group IIA secretory phospholipase A2 (sPLA(2)-IIA) has been studied extensively because of its involvement in inflammatory processes. Up-regulation of this enzyme has been shown in a number of neurodegenerative diseases including cerebral ischemia and Alzheimer's disease. PX-18 is a
Cristina Marcos-Arias et al.
BMC complementary and alternative medicine, 11, 119-119 (2011-11-29)
Candida-associated denture stomatitis is a frequent infectious disease. Treatment of this oral condition is difficult because failures and recurrences are common. The aim of this study was to test the in vitro antifungal activity of pure constituents of essentials oils.
Martin L Cross et al.
Clinical and vaccine immunology : CVI, 16(9), 1378-1380 (2009-07-03)
Cultures of Mycobacterium bovis BCG, comprising predominantly single-cell bacilli, were prepared in broth without animal-derived reagents. When formulated into a vegetable-derived lipid matrix, the vaccine was stable in vitro and was immunogenic in vivo upon feeding it to mice. This
Rokeya Tasneen et al.
Antimicrobial agents and chemotherapy, 52(10), 3664-3668 (2008-08-13)
PA-824 is in phase II clinical testing to treat tuberculosis. At a dose of 100 mg/kg of body weight, it has demonstrated bactericidal activity during the initial and continuation phases of treatment in a murine model of tuberculosis. In a

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service