Skip to Content
Merck
All Photos(1)

Key Documents

P5555

Supelco

Puerarin

analytical standard

Synonym(s):

8-(β-D-Glucopyranosyl-7-hydroxy-3- (4-hydroxyphenyl)-4H-1-benzopyran-4-one, Kakonein

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€96.00

€96.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€96.00

About This Item

Empirical Formula (Hill Notation):
C21H20O9
CAS Number:
Molecular Weight:
416.38
Beilstein:
64198
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€96.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

~80% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages
forensics and toxicology
veterinary

format

neat

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)ccc3C(=O)C(=COc23)c4ccc(O)cc4

InChI

1S/C21H20O9/c22-7-14-17(26)18(27)19(28)21(30-14)15-13(24)6-5-11-16(25)12(8-29-20(11)15)9-1-3-10(23)4-2-9/h1-6,8,14,17-19,21-24,26-28H,7H2/t14-,17-,18+,19-,21+/m1/s1

InChI key

HKEAFJYKMMKDOR-VPRICQMDSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Puerarin may be used as a reference standard for the determination of puerarin in rat plasma after administration of puerariae radix isoflavone, using high performance liquid chromatography (HPLC).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Lei Wang et al.
PloS one, 14(6), e0218490-e0218490 (2019-06-25)
Growing evidence indicates that the gut microbiota plays a significant role in the pathophysiological processes of obesity and its related metabolic symptoms in the host. Puerarin, an active ingredient in the root of Pueraria lobate has been suggested to have
HPLC method for the determination and pharmacokinetic studies on puerarin in cerebral ischemia reperfusion rat plasma after intravenous administration of puerariae radix isoflavone
Yan B, et al.
Journal of Pharmaceutical and Biomedical Analysis, 37(2), 297-301 (2005)
Benguo Liu et al.
Journal of agricultural and food chemistry, 60(51), 12501-12507 (2012-12-12)
Puerarin is a natural isoflavone, found in the Chinese medicinal plant Ge-gen, with many reported health-promoting properties. However, its low water solubility impedes its application in pharmaceutical and functional food products. This study explores the formation of inclusion complex between
Jia-Wei Wang et al.
The Journal of surgical research, 186(1), 328-337 (2013-10-02)
A wealth of evidence has suggested that oxidative stress is involved in the secondary brain injury after traumatic brain injury (TBI). Recently, numerous in vivo and in vitro studies were reported that puerarin could inhibit oxidative stress through the activation
Y Zou et al.
Free radical research, 47(1), 55-63 (2012-10-24)
Current evidence suggests that amyloid beta (Aβ) peptides may play a major role in the pathogenesis of Alzheimer's disease in part by eliciting oxidative stress. Puerarin, a major isoflavone glycoside from Kudzu root (Pueraria lobata), has been reported to exert

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service